Would you like to log in again?
MET (M1250T) Kinase Enzyme System, Promega
Supplier: Promega
V6448 V4168
PAV6448EA 8648.06 USD InStock
PAV6448 PAV4168
MET (M1250T) Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human MET (point mutant M1250T; amino acids 956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile MET (M1250T) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human MET (point mutant M1250T; amino acids 956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MET is a proto-oncogene that encodes a transmembrane growth factor receptor, which is a heterodimer of two disulphide-linked chains of 50kDa (alpha) and 145kDa (beta). MET is widely expressed in the kidney, brain, lung, skin and embryonic tissue. Hepatocyte growth factor (HGF) binds to MET and activates its tyrosine kinase activity. MET is overexpressed and activated in a variety of human cancers including pancreatic, colon, gastric, cervical and ovarian and has been shown to be involved in tumor cell migration and invasion. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: MET (M1250T), 10ug (Human, recombinant; amino acids 956-end). MW: ~81kDa. Substrate: Poly (4:1 Glu, Tyr) peptide. Other: Reaction Buffer, DTT. MnCl2. MET (M1250T) NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/4233/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty