Would you like to log in again?
MLK2 Kinase Enzyme System, Promega
Supplier: Promega
V4476 V6450
PAV4476EA 572.31 USD InStock
PAV4476 PAV6450
MLK2 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human MLK2 (1-446) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MLK2 or mixed-lineage kinase 2 is a member of the serine/threonine kinase family, which activates MAPK8/JNK1 and MKK4/SEK1.
  • Easily Screen and Profile MLK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human MLK2 (1-446) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MLK2 or mixed-lineage kinase 2 is a member of the serine/threonine kinase family, which activates MAPK8/JNK1 and MKK4/SEK1. MLK2 itself can be phoshorylated and activated by JNK kinases. MLK2 is also known as mitogen-activated protein kinase kinase kinase 10 (MAP3K10). MLK2 functions preferentially on the JNK signaling pathway, which is involved in nerve growth factor (NGF)-induced neuronal apoptosis. MLK2 was isolated as a cDNA fragment from MKN28 gastric cancer cell library using degenerate PCR (1). MLK2 is highly expressed in brain, skeletal muscle and testis (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: MLK2, 10ug (Human, recombinant; amino acids 1-446). MW: ~76kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. MLK2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/4294/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty