Would you like to log in again?
PIM1 Kinase Enzyme System, Promega
Supplier: Promega
V4032 V6471
PAV4032EA 572.21 USD InStock
PAV4032 PAV6471
PIM1 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human PIM1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PIM1 is a proto-oncogene that belongs to a family of serine/threonine protein kinases that are highly conserved in multicellular organisms.
  • Easily Screen and Profile PIM1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PIM1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PIM1 is a proto-oncogene that belongs to a family of serine/threonine protein kinases that are highly conserved throughout evolution in multicellular organisms. Originally identified from Moloney murine leukemia virus induced T-cell lymphomas in mice, PIM1 is involved in the control of cytokine-mediated cell proliferation, differentiation and survival of lymphoid and myeloid cells as well as others (1). Expression of PIM1 can be stimulated by a variety of growth factors and is regulated at four different levels: transcriptional, post-transcriptional, translational and post-translational (2). Expression of PIM1 is mediated through activation of the JAK/STAT pathway. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PIM1, 10ug (Human, recombinant full-length). MW: ~62kDa. Substrate: S6K Substrate (KRRRLASLR); derived from human 40S ribosomal protein S6 (amino acids 230-238). Other: Reaction Buffer, DTT. PIM1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5292/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty