Would you like to log in again?
PKC mu Kinase Enzyme System, Promega
Supplier: Promega
V4038 V6474
PAV6474EA 8821.02 USD InStock
PAV6474 PAV4038
PKC mu Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human PKC mu was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC mu is a member of the protein kinase C family that differs from the other PKC isoenzymes in structural and enzymatic properties.
  • Easily Screen and Profile PKCmicro Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PKC mu was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein Kinase C mu is a member of the protein kinase C (PKC) family that differs from the other PKC isoenzymes in structural and enzymatic properties. PKC mu is ubiquitous in nature with the highest expression in the thymus, lung and peripheral blood mononuclear cells (1). PKC mu forms a complex in vivo with a phosphatidylinositol 4-kinase and a phosphatidylinositol-4-phosphate 5-kinase. A region of PKC mu between the amino-terminal transmembrane domain and the pleckstrin homology domain is shown to be involved in the association with the lipid kinases (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKC mu, 10ug (Human, recombinant full-length). MW: ~131kDa. Substrate: CREBtide (KRREILSRRPSYR); derived from human CREB1 isoform A (amino acids 109-121). Other: Reaction Buffer, DTT. PKC mu NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5587/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty