Would you like to log in again?
MSK1 Kinase Enzyme System, Promega
Supplier: Promega
V5092 V6590
PAV6590EA 8648.06 USD InStock
PAV6590 PAV5092
MSK1 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human MSK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile MSK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human MSK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MSK1 or mitogen- and stress-activated protein kinase-1 contains two protein kinase domains and shares 43% protein sequence identity with the MAPKAPK1 isoforms. Northern blot analysis shows that MSK1 is expressed in all tissues with the highest level of expression in brain, muscle, and placenta. Immunoelectron microscopy localized MSK1 to the nucleus. MSK1 is activated in vitro and in vivo by either ERK or SAPK2 proteins. MSK1 rather than MAPKAP-K1 or MAPKAP-K2/K3, mediates activation of the cAMP response element-binding protein and activating transcription factor-1 by either growth factors or stress signals. By radiation hybrid analysis, the MSK1 gene has been mapped to chromosome 14q31-q32. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: MSK1, 10microg (Human, recombinant full-length). MW: ~120kDa. Substrate: RSK Substrate (KRRRLSSLRA); derived from human 40S ribosomal protein S6 (amino acid 230-239). Other: Reaction Buffer, DTT. MSK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/9252/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty