Would you like to log in again?
PKC beta I Kinase Enzyme System, Promega
Supplier: Promega
V6595 V5094
PAV6595EA 8821.02 USD InStock
PAV6595 PAV5094
PKC beta I Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human PKCbeta I was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKCβI is a member of the PKC family (phospholipid-dependent serine/threonine kinase) and is highly related to PKCβ II.
  • Easily Screen and Profile PKCβI Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PKCbeta I was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKCβI is a member of the PKC family (phospholipid-dependent serine/threonine kinase) and is highly related to PKCβ II. Unlike the mature PKCβ II mRNA and protein, which rapidly increase following acute insulin treatment, the PKCβI mRNA and protein levels remain unchanged. The stable overexpression of PKCβI, but not PKCβI, leads to insulin-stimulated glucose uptake into cells. Upon stimulation of B lymphocytes and mast cells, Syk regulates Btk, and Btk selectively regulates enzymatic activity of PKCβI. Specific regulation of PKCβI by Btk is consistent with the selective association of Btk with PKCβI. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKCβI, 10microg (Human, recombinant full length). MW: ~102kDa. Substrate: PKCtide (ERMRPRKRQGSVRRRV); derived from protein kinase C epsilon (amino acid 149-164). Other: Reaction Buffer, DTT, Lipid Activator Solution. PKCβI NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5579/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty