Would you like to log in again?
ADP-Glo Kinase Assay + ZAP70 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V8311
PAV8311EA 1220.65 USD InStock
PAV8311
ADP-Glo Kinase Assay + ZAP70 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human ZAP70 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ZAP70 is a nonreceptor protein tyrosine kinase (part of the Syk/Zap70 family) that is involved in signaling by the T-cell antigen receptor.
  • Easily Screen and Profile ZAP70 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human ZAP70 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ZAP70 is a nonreceptor protein tyrosine kinase (part of the Syk/Zap70 family) that is involved in signaling by the T-cell antigen receptor (TCR). Ligation of the TCR/CD3 receptor in Jurkat T-cells induces phosphoprotein complexes, which contain ZAP70. TCR zeta chains are initially phosphorylated by p56Lck that lead to the recruitment of ZAP70 via its SH2 domain. ZAP70 in turn phosphorylates other proteins in the TCR-phosphoprotein complex. One of the natural substrates for ZAP70 is the zeta-chain dimer of the TCR/CD3 complex. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: ZAP70, 10ug (Human, recombinant full-length). MW: ~96kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT, MnCl2. ZAP70 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/7535/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty