Would you like to log in again?
Luciferin Detection Reagent with esterase, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
V8930 V8931
PAV8930EA 142.41 USD InStock
PAV8930 PAV8931
Luciferin Detection Reagent with esterase, Promega
Cell Culture Supplements and Additives
A reagent to detect presence of pro-luciferin substrates used to monitor the activity of specific isoforms of cytochrome P450 or NAT2.
  • Reagent for Detection of Pro-Luciferin Substrates
  • Reagent available with, or without, esterase
  • Use in combination with the pro-luciferin substrates
A detection reagent for luciferin substrates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty