Would you like to log in again?
P450-Glo CYP3A4 Assay with Luciferin-IPA, Promega
Supplier: Promega
V9002 V9001
PAV9001EA 269.11 USD InStock
PAV9001 PAV9002
P450-Glo CYP3A4 Assay with Luciferin-IPA, Promega
Assays Cellular Assays
The P450-Glo CYP3A4 Assay System provides a homogeneous, luminescent method for measuring CYP3A4 activity and the screening system contains a complete set of reagents for performing the assay.
  • Luminescent Assays and Screening Systems for Measuring CYP3A4 Activity
  • Screening systems include complete set of reagents
  • Luminescent format eliminates need for time-consuming analyses
  • Broad dynamic range and low background
The P450-Glo CYP3A4 Assay System provides a homogeneous, luminescent method for measuring CYP3A4 activity. The assays are designed to measure the activities of P450s from recombinant and native sources and for testing the effects of analytes such as drugs and new chemical entities on P450 activities. These luminescent assays exhibit exquisite sensitivity, low background signals and broad dynamic range. P450-Glo Assays employ luminogenic P450 substrates that are derivatives of beetle luciferin, a substrate for luciferase enzymes. The derivatives are not substrates for luciferase but are converted by P450s to luciferin, which in turn reacts with luciferase to produce light that is directly proportional to the activity of the P450. The P450-Glo Assays generate a glow-type luminescent signal, produced using derivatized luciferins as P450 substrates and a recombinant stabilized luciferase (Ultra-Glo Luciferase) coupled with a proprietary buffer system. The half-life of the luminescent output is greater than two hours, eliminating the need for luminometers with injectors and allowing batch plate processing. The formulation also minimizes the incidence of false positives due to inhibition of luciferase by analytes when screening for cytochrome P450 inhibitors. Drug-induced changes in expression of CYP450 genes are a key cause of drug-drug interactions. The ability to measure enzymatic activity of the specific human isoforms that are induced is critical for developing safer drugs. Currently, the most important inducible human isoforms are CYP1A2, CYP2B6 and CYP3A4. With the introduction of the new P450-Glo CYP2B6 Assay we now have assays to measure the activities of all three of these important inducible isoforms. The luciferin-based substrates are readily taken up by cells and rapidly converted into luciferin inside the cell, which reduces the incubation time required (typically 30-60 minutes). The low background and high signal-to-noise ratios produced mean less starting material is required. Dimethyl sulfoxide (DMSO), a common solvent used to solubilize chemical compounds, can significantly inhibit the activity of the 3A4 isoform of cytochrome P450, even at low concentrations (<0.1%). The P450-Glo CYP3A4 System (Luciferin-PPXE) DMSO-Tolerant Assay is specifically designed to tolerate DMSO in the 3A4 reaction. The assay exhibits little to no change in the signal-to-background ratio in the presence of 0.2% DMSO compared to a no-DMSO control.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty