Would you like to log in again?
ADP-Glo Kinase Assay + AKT2 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9041
PAV9041EA 1196.72 USD InStock
PAV9041
ADP-Glo Kinase Assay + AKT2 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human AKT2 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. AKT2 or Protein Kinase B beta (PKB beta) is a serine/threonine kinase that is a member of the AKT family.
  • Easily Screen and Profile AKT2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human AKT2 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. AKT2 or Protein Kinase B beta (PKB beta) is a serine/threonine kinase that is a member of the AKT family. AKT2 like the other AKT members is activated in cells in response to diverse stimuli such as hormones, growth factors and extracellular matrix components and is involved in glucose metabolism, transcription, survival, cell proliferation, angiogenesis and cell motility. The PI3K generates phosphatidylinositol-3,4,5-trisphosphate (PIP3), a lipid second messenger essential for the translocation of AKT2 to the plasma membrane where it is phosphorylated and activated by phosphoinositide-dependent kinase-1 (PDK-1). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: AKT2, 10ug (Human, recombinant full-length). MW: ~85kDa. Substrate: Modified AKT Substrate peptide (modified CKRPRAASFAE); based on the N-terminus of GSK3. Other: Reaction Buffer, DTT. AKT2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/208/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty