Would you like to log in again?
ADP-Glo Kinase Assay + CAMK4 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9091
PAV9091EA 1196.72 USD InStock
PAV9091
ADP-Glo Kinase Assay + CAMK4 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human CAMK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMK4 is a multifunctional serine/threonine protein kinase and a member of Ca2+/calmodulin-dependent protein kinase family.
  • Easily Screen and Profile CAMK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human CAMK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMK4 is a multifunctional serine/threonine protein kinase and a member of Ca2+/calmodulin-dependent protein kinase family. CAMK4 is localized in neurons in the hippocampus, amygdala, anterior cingulate cortex, somatosensory cortex and insular cortex. CAMK4 is involved in neural activity-dependent signaling in the neuronal nucleus and thought to play an important role in the consolidation/retention of hippocampus-dependent long-term memory. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CAMK4, 10ug (Human, recombinant full-length). MW: ~79kDa. Substrate: Autocamtide-2 (KKALRRQETVDAL-amide); derived from the autophosphorylation site (amino acid 283-290) on CaMKII. Other: Reaction Buffer, DTT, Ca2+/Calmodulin solution. CAMK4 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/814/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty