Would you like to log in again?
ADP-Glo Kinase Assay + Aurora B Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9181
PAV9181EA 1196.72 USD InStock
PAV9181
ADP-Glo Kinase Assay + Aurora B Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human Aurora B was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. Aurora B is a member of the Aurora kinase family that associates with microtubules during chromosome movement and segregation.
  • Easily Screen and Profile Aurora B Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human Aurora B was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. Aurora B is a member of the Aurora kinase family that associates with microtubules during chromosome movement and segregation. Aurora B localizes to the microtubules near kinetochores, specifically to the specialized microtubules called K-fibers. Aurora B inhibits the microtubule depolymerizing activity of mitotic centromere-associated kinesin (MCAK) by phosphorylating MCAK on Ser92. This phosphorylation also regulates MCAK translocalization from kinetochores to the centromere. Aurora B has been identified as a target for the development of new anticancer agents since inhibition of Aurora B gives rise to the more pronounced antiproliferative phenotype. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: Aurora B, 10ug (Human, recombinant full-length). MW: ~68kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. Aurora B NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/9212/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty