Would you like to log in again?
ADP-Glo Kinase Assay + NEK2 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9231
PAV9231EA 1196.72 USD InStock
PAV9231
ADP-Glo Kinase Assay + NEK2 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human NEK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NEK2 is closely related in its catalytic domain to the serine/threonine protein kinase NIMA of Aspergillus nidulans.
  • Easily Screen and Profile NEK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human NEK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NEK2 is closely related in its catalytic domain to the serine/threonine protein kinase NIMA of Aspergillus nidulans that is required for entry into mitosis and may function in parallel to the universal mitotic inducer p34cdc2. Like NIMA, the NEK2 protein is almost undetectable during G1 but accumulated progressively throughout S, reaching maximal levels in late G2. NEK2 is shown to be expressed most abundantly in the testis of the adult tissues examined being localized to the nucleus. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: NEK2, 10ug (Human, recombinant full-length). MW: ~76kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. NEK2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/4751/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty