Would you like to log in again?
ADP-Glo Kinase Assay + CHK1 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9241
PAV9241EA 1220.65 USD InStock
PAV9241
ADP-Glo Kinase Assay + CHK1 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human CHK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CHK1 was originally identified in fission yeast to play a role in activation of the DNA damage checkpoint in the G2 phase of the cell cycle.
  • Easily Screen and Profile CHK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human CHK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CHK1 is a 56kDa serine/threonine protein kinase that was originally identified in fission yeast to play a role in activation of the DNA damage checkpoint in the G2 phase of the cell cycle. CHK1 appears to function downstream of several of the known fission yeast checkpoint gene products, including that encoded by rad3+, a gene with sequence similarity to the ATM gene mutated in patients with ataxia telangiectasia. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CHK1, 10ug (Human, recombinant full-length). MW: ~82kDa. Substrate: CHKtide (KKKVSRSGLYRSPSMPENLNRPR); derived from the human CDC25C protein isoform A (amino acid 205-225). Other: Reaction Buffer, DTT. CHK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1111/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty