Would you like to log in again?
ADP-Glo Kinase Assay + FYN A Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9341
PAV9341EA 1266.1 USD InStock
PAV9341
ADP-Glo Kinase Assay + FYN A Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Full-length recombinant human FYN A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FYN A (Fyn isoform A) is a member of the SRC tyrosine kinase oncogene family showing high homology to YES1, FGR and SRC.
  • Easily Screen and Profile FYN A Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Full-length recombinant human FYN A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FYN A (Fyn isoform A) is a member of the SRC tyrosine kinase oncogene family showing high homology to YES1, FGR and SRC. FYN has been shown to phosphorylate DAB1, an intracellular adaptor protein that interacts with amyloid precursor protein (APP) and apoE receptor 2 (apoEr2). The interaction of FYN and DAB1 regulates the phosphorylation, trafficking and processing of APP and apoEr2. FYN expression has been shown to be significantly increased in Chronic Myelogenous Leukemia (CML). Knockdown of FYN with shRNA slows leukemia cell growth, inhibits clonogenicity and leads to increased sensitivity to imatinib. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: FYN A, 10ug (Human, recombinant full-length). MW: ~85kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT. FYN A NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/2534/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty