Would you like to log in again?
ADP-Glo Kinase Assay + KDR Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9471
PAV9471EA 1196.72 USD InStock
PAV9471
ADP-Glo Kinase Assay + KDR Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human KDR (amino acids 789-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. KDR (or kinase insert domain receptor) is a growth factor receptor tyrosine kinase originally isolated from human endothelial cells.
  • Easily Screen and Profile KDR Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human KDR (amino acids 789-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. KDR (or kinase insert domain receptor) is a growth factor receptor tyrosine kinase that was originally isolated from human endothelial cells where it plays a pivotal role in endothelial cell proliferation and differentiation. KDR and its mouse homolog Flk1 bind VEGF with high affinity and are implicated in the development of new blood vessels (angiogenesis). The expression levels of VEGF and KDR are highly correlated during the normal development of the ocular vasculature in humans. Induction of angiogenesis is a critical step in tumor progression, and inhibitors of KDR have been demonstrated both to induce tumor regression and reduce metastatic potential in preclinical models. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: KDR, 10ug (Human, recombinant; amino acids 789-end). MW: ~110kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT. KDR NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/3791/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty