Would you like to log in again?
ADP-Glo Kinase Assay + ASK1 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9481
PAV9481EA 1196.72 USD InStock
PAV9481
ADP-Glo Kinase Assay + ASK1 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human ASK1 (649-946) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. ASK1, also known as MAPKKK5, activates MKK3, MKK4 (SEK1) and MKK6. Overexpression of ASK1 induces apoptotic cell death.
  • Easily Screen and Profile ASK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human ASK1 (649-946) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. ASK1, also known as MAPKKK5, activates MKK3, MKK4 (SEK1) and MKK6. Overexpression of ASK1 induces apoptotic cell death, and ASK1 is activated in cells treated with tumor necrosis factor-alpha. ASK1 interacts with members of the TRAF family and is activated by TRAF2 in the TNF-signaling pathway. After activation by TRAF2, ASK1 activates MKK4, which in turn activates JNK. Thus, ASK1 is a mediator of TRAF2-induced JNK activation. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: ASK1, 10ug (Human, recombinant; amino acids 649-946). MW: ~60kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. ASK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/4217/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty