Would you like to log in again?
ADP-Glo Kinase Assay + SGK1 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9671
PAV9671EA 1266.1 USD InStock
PAV9671
ADP-Glo Kinase Assay + SGK1 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human SGK1 (amino acids 60-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SGK1 is a member of the serum- and glucocorticoid-induced protein kinase family.
  • Easily Screen and Profile SGK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human SGK1 (amino acids 60-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SGK1 is a member of the serum- and glucocorticoid-induced protein kinase family that is activated in vitro by 3-phosphoinositide-dependent protein kinase-1 (PDK1) and in vivo in response to signals that activate phosphatidylinositol (PI) 3-kinase. SGK1 mRNA is expressed in all tissues, and the level of SGK1 mRNA is increased by stimulation with serum or dexamethasone. SGK1 promotes cell survival by phosphorylating and inactivating FKHRL1. SGK and Akt display differences with respect to the efficacy with which they phosphorylate the three regulatory sites on FKHRL1. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: SGK1, 10ug (Human, recombinant; amino acids 60-end). MW: ~73kDa. Substrate: Akt (PKB) substrate (CKRPRAASFAE). Other: Reaction Buffer, DTT. SGK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/6446/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty