Would you like to log in again?
ADP-Glo Kinase Assay + PKC alpha Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9691
PAV9691EA 1196.72 USD InStock
PAV9691
ADP-Glo Kinase Assay + PKC alpha Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human PKC alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC alpha is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases.
  • Easily Screen and Profile PKCα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PKC alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC alpha is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC alpha is reported to play roles in many different cellular processes, such as cell adhesion, cell transformation, cell cycle checkpoint and cell volume control. PKC alpha has been assigned to the chromosome region 17q22-q23.2 and has been identified as a fundamental regulator of cardiac contractility and Ca2+ handling in myocytes. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKC alpha, 10ug (Human, recombinant full-length). MW: ~103kDa. Substrate: CREBtide (KRREILSRRPSYR); derived from human CREB1 isoform A (amino acid 109-121). Other: Reaction Buffer, DTT, Lipid solution. PKC alpha NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5578/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty