Would you like to log in again?
ADP-Glo Kinase Assay + PKC gamma Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9711
PAV9711EA 1220.65 USD InStock
PAV9711
ADP-Glo Kinase Assay + PKC gamma Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human PKC gamma was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC gamma is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases.
  • Easily Screen and Profile PKCγ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PKC gamma was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC gamma is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases that can phosphorylate a wide variety of protein targets known to be involved in diverse cellular signaling pathways. In the brain, PKC gamma is translocated to cell membranes during ischemia and is rapidly removed or degraded during the second otherwise lethal ischemic insult in preconditioned brains. This suggests that ischemic preconditioning enhances downregulation of cell signaling mediated by PKC gamma and may thereby provide neuroprotection. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKC gamma, 10ug (Human, recombinant full-length). MW: ~105kDa. Substrate: PKCtide (ERMRPRKRQGSVRRRV); derived from protein kinase C epsilon (amino acid 149-164). Other: Reaction Buffer, DTT, Lipid solution. PKC gamma NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5582/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty