Would you like to log in again?
ADP-Glo Kinase Assay + PKC delta Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9721
PAV9721EA 1220.65 USD InStock
PAV9721
ADP-Glo Kinase Assay + PKC delta Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human PKC delta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein kinase C delta is a member of the protein kinase C (PKC) family of serine-threonine kinases.
  • Easily Screen and Profile PKCδ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PKC delta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein kinase C delta is a member of the protein kinase C (PKC) family of serine-threonine kinases. It is a 79kDa protein kinase that shows strict dependence on the presence of phospholipids but shows no activation by Ca2+. Phosphatidylinositol is the most potent activator of PKC delta. Northern blot analysis indicated that PKC delta is widely distributed in almost all tissues and is a major isoform of PKC expressed in hemopoietic cells. PKC delta is involved in fundamental cellular functions regulated by diacylglycerols and mimicked by phorbol esters. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKC delta, 10ug (Human, recombinant full-length). MW: ~104kDa. Substrate: CREBtide (KRREILSRRPSYR); derived from human CREB1 isoform A (amino acid 109-121). Other: Reaction Buffer, DTT, Lipid solution. PKC delta NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5580/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty