Would you like to log in again?
ADP-Glo Kinase Assay + SRC Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9741
PAV9741EA 1196.72 USD InStock
PAV9741
ADP-Glo Kinase Assay + SRC Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human SRC was expressed in E. coli cells using an N-terminal GST tag. SRC was originally identified as a transforming protein of the Rous sarcoma virus that had enzymatic ability to phosphorylate tyrosine in protein substrates.
  • Easily Screen and Profile SRC Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human SRC was expressed in E. coli cells using an N-terminal GST tag. SRC family belongs to nonreceptor tyrosine kinases. SRC was originally identified as a transforming protein of the Rous sarcoma virus (RSV) that had enzymatic ability to phosphorylate tyrosine in protein substrates. SRC is overexpressed and activated in a large number of human malignancies and has been linked to the development of cancer and progression to distant metastases. In addition to increasing cell proliferation, a key role of SRC in cancer seems to be the ability to promote invasion and motility, functions that might contribute to tumor progression. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: SRC, 10ug (Human, recombinant full-length). MW: ~83kDa. Substrate: SRC substrate (KVEKIGEGTYGVVYK-amide); derived from human p34cdc2 (amino acids 6-20). Other: Reaction Buffer, DTT, MnCl2. SRC NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/6714/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty