Would you like to log in again?
SV Total RNA Isolation System, Promega
Supplier: Promega
Z3100 Z3101 Z3105
PAZ3101EA 147.28 USD InStock
PAZ3101 PAZ3105 PAZ3100
SV Total RNA Isolation System, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
A fast, simple technique for preparation of intact total RNA from tissues, cultured cells and white blood cells in as little as one hour.
  • Extracts RNA from Tissues, Cultured Cells and White Blood Cells
  • Rapid isolation of high yields of total RNA without the use of hazardous compounds
  • Choice of microcentrifugation (spin) or vacuum purification methods
  • Purified RNA suitable for all routine molecular biology applications, including RT-PCR
The SV Total RNA Isolation System provides a fast and simple technique for preparation of intact total RNA from tissues, cultured cells and white blood cells in as little as one hour. Using this membrane-based purification system, up to 60mg of tissue can be processed per purification, depending on tissue type. The system incorporates a DNase treatment step directly on the minicolumn membrane. This step substantially reduces genomic DNA contamination, which can interfere with amplification-based methodologies. Purification is achieved without the use of phenol:chloroform extractions or ethanol precipitations, and there is no DNase carryover in the final RNA preparation.
Purification is achieved without the use of phenol: chloroform extractions or ethanol precipitations, and there is no DNase carryover in the final RNA preparation.
The system incorporates a DNase treatment step directly on the minicolumn membrane. This step substantially reduces genomic DNA contamination, which can interfere with amplification-based methodologies.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty