Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MagneSphere Technology Magnetic Separation Stand (two-position), Promega
Supplier: Promega
Z5331 Z5332 Z5333
PAZ5332EA 66.64 USD InStock
PAZ5332 PAZ5333 PAZ5331
MagneSphere Technology Magnetic Separation Stand (two-position), Promega
Racks Magnetic Separation Racks
Use in conjunction with the PolyATtract Systems. Ideal for applications requiring multiple paramagnetic isolations of biomolecules.
  • Use with both MagneSphere(R) and PolyATtract(R) System components
  • Conduct simultaneous processing of the contents of two tubes or twelve tubes of various sizes
The MagneSphere Technology Magnetic Separation Stands can be used in conjunction with any of the PolyATtract Systems and are ideal for applications requiring multiple paramagnetic isolations of biomolecules. These stands use the same strong rare earth magnet used in the PolyATtract Systems Magnetic Separation Stands and come in a variety of sizes to accommodate 2-96 samples.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty