Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PolyATtract System 1000 with Magnetic Stand, Scalable, Promega

Supplier: Promega

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Z5420 Z5400
PAZ5400EA 725.06 USD InStock
PAZ5400 PAZ5420
PolyATtract System 1000 with Magnetic Stand, Scalable, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The PolyATtract System 1000 isolates messenger RNA directly from crude cell or tissue lysates, eliminating the need for total RNA isolation.

  • Isolates mRNA Directly from Crude Cell or Tissue Lysates
  • mRNA Purification is complete 45 minutes or less
  • Works with tissue amounts from 5mg-2g per isolation
  • No lengthy ethanol precipitation steps, phenol:chloroform extractions, and overnight ultracentrifugation
  • Magnetic separation stand (Cat.# Z5410) accommodates 1.5ml, 2ml, 15ml and 50ml tube sizes

The PolyATtract System 1000 isolates messenger RNA directly from crude cell or tissue lysates, eliminating the need for total RNA isolations. This system uses the MagneSphere technology for the purification of poly(A)+ RNA, eliminating the need for oligo(dT) cellulose columns. The increased yield of mRNA using this system allows the detection of low-copy-number mRNAs in relatively small amounts of material using Northern blot analysis. The isolated mRNA is suitable for all molecular biology applications, including in vitro translation, cDNA synthesis, PCR analysis, ribonuclease (RNase) protection assays, primer extension and Northern blots. The MagneSphere Technology Magnetic Separation Stands can be used in conjunction with any of the PolyATtract Systems and are ideal for applications requiring multiple paramagnetic isolations of biomolecules.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty