Would you like to log in again?
RNAgents® Denaturing Solution, Promega
Supplier: Promega
Z5651
PAZ5651EA 188.02 USD InStock
PAZ5651
RNAgents® Denaturing Solution, Promega
Buffers Extraction Buffers
RNAgents Denaturing Solution lyses cells or tissue under conditions that rapidly inhibit ribonucleases.
  • Lyses Cells or Tissue Under Conditions that Rapidly Inhibit Ribonucleases
  • Uses two potent inhibitors of RNase, guanidine thiocyanate and β-mercaptoethanol
  • Designed to be used with acidic phenol:chloroform and alcohol (isopropanol) for purification of total RNA
  • Fits RNAgents® Total RNA Isolation System (PAZ5110)
RNAgents Denaturing Solution lyses cells or tissue under conditions that rapidly inhibit ribonucleases, using two potent inhibitors of RNase, guanidine thiocyanate and beta-mercaptoethanol. The RNAgents Denaturing Solution is designed to be used in concert with acidic phenol:chloroform and alcohol (isopropanol) for purification of total RNA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty