Would you like to log in again?
Amniocentesis OB/GYN Tray, BD
Supplier: BD Medical
4545A
PH-4545ACS 227.97 USD InStock
PH-4545A
Amniocentesis OB/GYN Tray, BD
Trays/Dishes/Pots Instrument Trays/Dishes
These trays support amniocentesis, paracervical/pudendal nerve blocks and circumcision procedures.
  • For preventable medical error reduction
  • Sharpens safety
  • Infection prevention
  • Single use
Delivery information: Includes: Spinal needle, 20 G×3.5", centimeter depth markings; Lock-tip syringes,5 and 20ml; Lidocaine hydrochloride,USP,1%,5 ml; three amber specimen vials,10 ml; Prep cup; three swab sticks; Ruler,15 cm; Three gauze pads; Towel; Fenestrated drape; Bandage; Hospital wrap.
Packaging: 10/case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty