Would you like to log in again?
CP100™ BT Sterile Latex Hand-Specific Gloves, Cardinal
Supplier: Cardinal
CP100™
2Y5003 2Y5000 2Y5005 2Y5004 2Y5001 2Y5007 2Y5002 2Y5000 2Y5006
PH2Y5005CS 1768.3 USD InStock
PH2Y5005 CAPH2Y5000 PH2Y5004 PH2Y5003 PH2Y5002 PH2Y5001 PH2Y5000 PH2Y5007 PH2Y5006
CP100™ BT Sterile Latex Hand-Specific Gloves, Cardinal
Gloves
Sterile, clean-processed gloves feature a comfortable flex-and-stretch design and low particulate levels for minimized contamination of controlled environments.
  • Ambidextrous, beaded cuff
  • Fully textured palm and fingers provide a secure grip
  • Clean processed, powder free
The antislip, textured finish enhances the grip for better handling. Tapered fingers help reduce fatigue while providing increased tactile sensitivity. Beaded cuff with reinforced ribs prevents constriction and reduces rolldown. Hand-specific. Powder-free. Color: natural white.
Certifications: Sterilized by gamma irradiation to a Sterility Assurance Level of 10⁻⁶ in accordance with AAMI guidelines. Certificates of sterility available upon request. Each lot is performance tested and is fully traceable.
Ordering information: 50 individually packed pairs per poly bag, four poly bags per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty