Would you like to log in again?
Pierce™ Active GTPase Pull-down and Detection Kit, Thermo Scientific
Supplier: Thermo Scientific
Pierce
16118 16117 16118 16116 16120 16117 16119 16119 16116 16120 16121 16121
PI16120EA 1287.14 USD InStock
PI16120 CAPI16121 PI16121 CAPI16120 CAPI16119 PI16116 CAPI16118 PI16117 PI16118 PI16119 CAPI16117 CAPI16116
Pierce™ Active GTPase Pull-down and Detection Kit, Thermo Scientific
Assays Enzyme Assays
The Thermo Scientific Active GTPase Pull-Down and Detection Kits allows the monitoring of changes in GTPase activity levels by enriching and detecting the active (GTP-bound) state of a particular GTPase.
  • Validated – functionally tested to ensure quality and performance
  • Sensitive – optimised antibodies, reagents and Western blotting procedure accurately detect changes in GTPase activity levels
  • Convenient – kits contain controls and all reagents needed to perform and detect 30 pull-downs
  • Easy to use – conditions are optimised for immediate success in a 2-hour assay
  • Efficient – spin columns prevent sample loss
These kits include all the necessary reagents to lyse cells, enrich for active GTPases, positive and negative controls and an anti-GTPase primary antibody for Western blot detection.
Delivery information: Each kit includes GST fusion protein of specific binding domain (1 vial), glutathione agarose resin (3 ml), GTPγS, 100X (50 µl), GDP 100X (50 µl), lysis/binding/wash buffer (100 ml), GTPase-specific primary antibody (1 vial), SDS-PAGE sample loading buffer, 2X (1,5 ml), spin columns (30), and collection tubes (90).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty