Would you like to log in again?
EZ-Link™ Desthiobiotinylation and Pull-Down Kit and Reagents, Thermo Scientific
Supplier: Thermo Scientific
Pierce
16130 16138 16138 16130
PI16130EA 538.54 USD InStock
PI16130 CAPI16130 PI16138 CAPI16138
EZ-Link™ Desthiobiotinylation and Pull-Down Kit and Reagents, Thermo Scientific
Protein Linking and Labeling Systems
Thermo Scientific EZ-Link™ Desthiobiotinylation and Pull-down Kit is a complete set of reagents to biotin-label purified proteins and use them to purify interaction complexes using streptavidin agarose resin with mild elution conditions.
  • Sulpho-NHS-LC-Desthiobiotin label proteins with a biotin-like that specifically binds streptavidin
  • Simple, optimized protocol and reagents to label proteins
  • Recover (purify) intact, functional protein interaction complexes
  • PEG groups in spacer arms enhance water solubility of biotinylated molecules and help prevent aggregation of biotinylated antibodies
EZ-Link™ Desthiobiotinylation and Pull-down Kit uses Sulpho-NHSLC-Desthiobiotin reagents to label target proteins on primary amines, desalting columns to clean up the labeling reaction, and immobilized high capacity streptavidin resin and buffers to perform pull-down assays
Pull-down assays use the tagged or labelled bait protein coupled to resin to capture a prey protein contained in a cell lysate or other unpurified protein mixtures. This technique can identify novel interactions between a known protein (bait) and previously undiscovered target (prey) or it can confirm the interaction between a known (bait) and a known target (prey) protein.
Delivery information: EZ-Link™ Desthiobiotinylation and Pull-down Kit includes EZ-Link Sulpho-NHS-LC-Desthiobiotin (2×1 mg), Zeba™ spin desalting column, 7K MWCO, 5 mL (5 columns), high capacity streptavidin agarose resin (2 mL), PBS (1 pack, makes 500 mL), Biotin elution buffer (8 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty