Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ Gaussia Luciferase Glow Assay Kit, Thermo Scientific

Supplier: Thermo Scientific
Pierce™
16161 16160 16160 16161
PI16160EA 157.2 USD InStock
CAPI16161 CAPI16160 PI16160 PI16161
Pierce™ Gaussia Luciferase Glow Assay Kit, Thermo Scientific
Assays Enzyme Assays
The Thermo Scientific Pierce Gaussia luciferase glow assay kit provides an extremely bright and stable bioluminescence signal (half-life greater than 1 hour), especially in the presence of Gaussia-Dura luciferase reporter.

  • Highly sensitive detection of Gaussia luciferase activity
  • Increased signal stability
  • Assay reagents compatible with other Gaussia luciferases
  • Automation-friendly - amenable to high throughput assays
  • Contains a universal cell lysis buffer and optimized glow assay reagent
  • Allows one to perform non-radioactive assays

Light output generated by the luciferase reaction can be correlated to the amount of Gaussia luciferase protein produced and used to determine the activity of the promoter driving Gaussia expression

Kit may be stored at –80 ˚C, or individually at indicated temperatures.

The Gaussia Glow Assay Kit reagent yields a Gaussia-Dura luciferase reaction with glow-type kinetics and is recommended for luminometers without injectors or for applications requiring batch processing of samples.

The signal produced by Gaussia luciferase shows strong flash kinetics and is considerably greater than flash signals from either firefly or Renilla luciferases assayed under similar conditions.

Delivery information: Kit contains Gaussia glow assay buffer (5 or 50 ml) (store at 4 ˚C), 100X Coelenterazine (50 or 500 µl) (store at –80 ˚C) and 2X cell lysis buffer (6 or 60 ml) (store at room temperature).

Caution: For research use only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty