Would you like to log in again?
Pierce™ Renilla Luciferase Flash Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
Pierce™
16164 16165 16165 16164
PI16164EA 135.6 USD InStock
PI16164 CAPI16165 PI16165 CAPI16164
Pierce™ Renilla Luciferase Flash Assay Kit, Thermo Scientific
Assays Enzyme Assays
The Thermo Scientific Pierce Renilla luciferase flash assay kit provides researchers with a highly sensitive intracellular assay for transcriptional activity of regulatory elements in mammalian whole cell lysate.
  • Greater sensitivity allows utilization of smaller numbers of cells
  • Highest sensitivity assays result in decreased reagent consumption
  • Assays require minimal sample handling
  • Amenable to automation
  • Contains a universal cell lysis buffer and optimized flash assay reagent
  • Allows one to perform non-radioactive assays
This Flash Assay Kit contains reagents for measuring the activity of Renilla luciferase in mammalian cell lysates.
The protein provides extremely bright flash signal that decays rapidly. The Pierce Renilla Flash Assay Kit reagent optimizes the this signal for use in flash-type luciferase reporter assays. The kit also is compatible for flash assays with other Renilla luciferases and derivatives that use coelenterazine as a substrate.
When used with Thermo Scientific Green Renilla Luc Plasmids, the kit provides an extremely sensitive bioluminescent reporter assay system for intracellular detection of promoter or pathway activity. The signal produced by Green Renilla Luc is considerably greater than signal from either native firefly or Renilla luciferases assayed under similar conditions. Light output captured using a luminometer can be correlated with the amount of Renilla luciferase protein produced and used to determine the activity of the promoter driving Renilla expression.
Delivery information: Kit contains Renilla flash assay buffer(5 or 50 ml) (store at 4 ˚C), 100X Coelenterazine (50 or 500 µl) (store at –80 ˚C) and 2X cell lysis buffer (6 or 60 ml) store at room temperature.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty