Would you like to log in again?
Pierce™ Renilla Luciferase Glow Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
Pierce
16166 16166 16167 16167
PI16166EA 156 USD InStock
CAPI16166 PI16166 CAPI16167 PI16167
Pierce™ Renilla Luciferase Glow Assay Kit, Thermo Scientific
Assays Enzyme Assays
The Thermo Scientific Pierce Renilla Luciferase Glow Assay Kit provides an extremely bright and stable bioluminescence signal (half-life approx
  • Sensitive—highly sensitive detection of Green Renilla luciferase activity
  • Stable—increased signal stability compared to flash assays
  • Compatible—assay reagents compatible with other Renilla luciferases
  • Automation-friendly—amenable to high throughput assays
  • Convenient—contains a universal cell lysis buffer and optimized glow assay reagent
3 hours), especially in the presence of Green Renilla luciferase reporter.
Delivery information: Kit contains Renilla Glow Assay Buffer (5 mL); 100X Coelenterazine (50 µL); and 2X Cell Lysis Buffer (6 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty