Would you like to log in again?
Pierce™ Cypridina Luciferase Flash Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
Pierce™
16168 16168 16169 16169
PI16168EA 140.42 USD InStock
CAPI16168 PI16168 PI16169 CAPI16169
Pierce™ Cypridina Luciferase Flash Assay Kit, Thermo Scientific
Assays Enzyme Assays
The Thermo Scientific Pierce Cypridina Luciferase Flash Assay Kit provides researchers with a highly sensitive assay for transcriptional activity of regulatory elements in mammalian cell culture media and whole cell lysate.
  • Sensitive—high sensitivity allows utilization of smaller numbers of cells
  • Secreted luciferase—allows real-time assays and kinetic studies without destroying cells
  • Ideal for co-transfection—uses a different substrate than Renilla, Gaussia, or firefly luciferases
  • Compatible—assay reagents compatible with other Cypridina or Vargula luciferases
  • Automation-friendly—amenable to high throughput screens
  • Convenient—contains a universal cell lysis buffer and optimized flash assay reagent
This Flash Assay Kit contains reagents for measuring the activity of Cypridina luciferase in mammalian cell culture media and lysates. Cypridina luciferase is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. The signal produced by Cypridina is considerably greater than signal from either firefly or Renilla luciferases assayed under similar conditions.
Delivery information: Kit includes Cypridina flash assay buffer (5 ml), 1X Vargulin (5 µl) and 2X Cell Lysis Buffer (6 ml).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty