Would you like to log in again?
Pierce™ Firefly Luciferase Flash Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
Pierce™
16175 16174 16175 16174
PI16174EA 136.82 USD InStock
PI16174 PI16175 CAPI16175 CAPI16174
Pierce™ Firefly Luciferase Flash Assay Kit, Thermo Scientific
Assays Protein Assays
The Pierce™ Firefly Luciferase Flash Assay Kit provides reagents to measure Red firefly luciferase activity in whole cell producing a very strong flash signal which decays rapidly.
  • Highly sensitive detection of firefly luciferase activity
  • Amenable to automation
  • Contains a universal cell lysis buffer and optimized flash assay reagent
The bioluminescent signal produced by firefly luciferase results from the oxidation of D-Luciferin. The light output correlates with the amount of firefly protein expressed, which is proportional to the activity of the promoter for firefly expression.
Delivery information: Kit contains Firefly flash assay buffer (5 or 50 ml), D-Luciferin (3 or 30 mg) and 2X Cell lysis buffer (6 or 60 ml).
Caution: For research use only. Not for use in diagnostic procedures. It stored at 4 ℃.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty