Would you like to log in again?
Pierce™ Luciferase Cell Lysis Buffer (2X), Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Pierce Pierce™
16189 16189
PI16189EA 296.4 USD InStock
CAPI16189 PI16189
Pierce™ Luciferase Cell Lysis Buffer (2X), Thermo Scientific
Buffers Extraction Buffers
The Thermo Scientific Pierce Luciferase Cell Lysis Buffer effectively lyses adherent cells or pelleted suspension cells for analysis of promoter expression in luciferase assays.
  • Compatible with all Pierce Luciferase Reporter Assays
  • Enables rapid and efficient lysis
  • Tested compatibility with multiple Mammalian cell lines
  • Compatible with protein assays (e.g., Coomassie Plus and BCA)
Pierce Luciferase Cell Lysis Buffer has been tested and validated to produce luciferase assay-compatible lysates from a variety of cell types. Essentially complete lysis of adherent cells has been observed with nearly all of the cell types tested to date, including COS-7, NIH3T3, Hepa 1-6, 293, CHO, MDA, MB 231, Jurkat, and FM2 cells.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty