Would you like to log in again?
Pierce™ Firefly Luc One-Step Glow Assay Kit
Supplier: Thermo Scientific
Pierce™
16197 16196 16196 16197
PI16196EA 202.8 USD InStock
PI16196 PI16197 CAPI16197 CAPI16196
Pierce™ Firefly Luc One-Step Glow Assay Kit
Assays Enzyme Assays
The Thermo Scientific Pierce Firefly One-Step Luc Glow Assay Kit measures firefly luciferase activity in mammalian cell culture using a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications.
  • Sensitive—highly sensitive detection of firefly luciferase activity
  • Odorless—reagents have no foul or toxic odor, eliminating the need to work in a fume hood
  • Convenient—one-step, mix-and-read protocol; no additional reagents required
  • Stable—increased signal half-life and stability compared to flash assays
  • Simple—does not require a luminometer with injectors
  • Automation-friendly—amenable to high throughput assays
Firefly luciferase bioluminescence provides researchers with a non-radioactive assay for measuring transcriptional activity of regulatory elements. Firefly luciferase reporter activity can be used to determine promoter activity, to study upstream pathway activity in the cell, or to determine effect of activators or inhibitors.
Delivery information: Kit includes Firefly Luc One-Step Substrate, lyophilised (makes 1 µl of 1X); Firefly Luc One-Step Assay Buffer (11 ml).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty