Would you like to log in again?
Pierce™ RNA 3' End Desthiobiotinylation Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
20163 20163
PI20163EA 853.54 USD InStock
CAPI20163 PI20163
Pierce™ RNA 3' End Desthiobiotinylation Kit, Thermo Scientific
Protein Linking and Labeling Systems
The Thermo Scientific Pierce RNA 3'-End Desthiobiotinylation Kit contains reagents for easy and efficient desthiobiotin labeling of RNA for use as probes or as antibody alternatives for enrichment of RNA binding proteins.
  • Non-radioactive— a single biotin labels has similar detection sensitivity as radioactivity
  • End-labeled—results in minimal disturbance of RNA secondary structure
  • Flexible—desthiobiotin label may be used for detection or as an affinity handle for streptavidin resin
The Pierce RNA 3'-End Desthiobiotinylation Kit is optimized for labeling the 3'-end of single-stranded RNA using T4 RNA ligase. Once labeled, the RNA can be used as a probe or target for gel-shift EMSA reactions, northern blots and protein-RNA interaction experiments. The desthiobiotinylated RNA also can be used to enrich for RNA binding proteins (RBP) using streptavidin affinity resin.
Delivery information: Kit includes desthiobiotinylated cytidine bisphosphate (40 µL), T4 RNA ligase (40 µL), T4 RNA ligase reaction buffer,10X (100 µL), Non-labeled RNA control (100 µL), Biotinylated IRE RNA control (35 µL), RNase inhibitor (2x10 µL), DMSO (200 µL), PEG 30% (300 µL), Glycogen (20 µL) and Nuclease-free water (1,5 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty