Would you like to log in again?
Pierce™ Immobilized GST Agarose, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
20211 20211
PI20211EA 899.45 USD InStock
PI20211 CAPI20211
Pierce™ Immobilized GST Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Immobilized GST is glutathione S-transferase from Schistosoma japonicum that has been covalently immobilized on crosslinked, 6% beaded agarose.
  • Provides purification of specific antibodies from those directed against the GST tag
  • Bound antibodies can be eluted with 6 M guanidinium chloride allowing for 10 purifications with the support
Antibody molecules recognizing the GST portion of a recombinant fusion protein can be removed from the desired antibody by passing antiserum over an immobilized GST column
The expression of foreign proteins in E. coli using a glutathione S-transferase (GST) fusion system is a method of choice for studying the function of particular proteins, for purifying interacting factors and for generating antibodies against fused proteins.
GST antibodies will bind to the immobilized GST column, whereas antibodies against other fusion proteins will pass through the column unrestrained.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty