Would you like to log in again?
Bovine Tpck Trypsin (from Pancreas)
Supplier: Pierce Chemical
Pierce
20233 20233
PI20233EA 146.83 USD InStock
PI20233 CAPI20233
Bovine Tpck Trypsin (from Pancreas)
Enzymes
Thermo Scientific Pierce TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.
  • Cleaves at carboxyl side of arginine and lysine residues
  • Digestion conditions: pH 7.5-9.0, 37°
Immobilized TPCK Trypsin can be substituted for free trypsin in many application and is advantageous because it minimizes autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the trypsin. Immobilized trypsin is also more stable against heat-induced denaturation, resulting in longer maintenance of activity.
Ordering information: Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8° C.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty