Would you like to log in again?
Pierce™ GST, Pierce Chemical
Supplier: Pierce Chemical
Pierce
20237 20237
PI20237EA 438.09 USD InStock
CAPI20237 PI20237
Pierce™ GST, Pierce Chemical
Proteins and Peptides
Glutathione S-transferase (GST) is a 211 amino acid protein (26kDa) whose DNA sequence is frequently integrated into expression vectors for production of recombinant proteins.
  • Solution of glutathione-S-transferase (≥100 units/mg)
  • Highly soluble, stable protein
  • GST-tagged fusion proteins can be purified or detected based on the ability of GST
The result of expression from this vector is a GST-tagged fusion protein in which the functional GST protein (26kDa) is fused to the N-terminus of the recombinant protein.
Because GST rapidly folds into a stable and highly soluble protein upon translation, inclusion of the GST tag often promotes greater expression and solubility of recombinant proteins than expression without the tag. In addition, GST-tagged fusion proteins can be purified or detected based on the ability of GST (an enzyme) to bind its substrate, glutathione (GSH).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty