Would you like to log in again?
CarboxyLink™ Coupling Resin, Thermo Scientific
Supplier: Thermo Scientific
CarboxyLink™
20266 20266
PI20266EA 308.86 USD InStock
PI20266 CAPI20266
CarboxyLink™ Coupling Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific CarboxyLink Coupling Resin is for covalent immobilization of peptides or other carboxyl-containing (—COOH) molecules to a porous, beaded resin for use in affinity purification procedures.
  • DADPA-activated crosslinked 4% beaded agarose
  • Couple 1 to 2 mg of peptide per mL of resin
  • Immobilizes peptides with high capacity and various orientations without steric hindrance
  • Immoblization results in covalent attachment of carboxyl groups by amide bonds, allowing for multiple rounds of affinity purification with one batch of prepared resin
CarboxyLink Resin is crosslinked beaded agarose that has been activated with diamino-dipropylamine (DADPA) to contain long spacer arms, each with a primary amine at the end. When incubated with the resin and the carbodiimide crosslinker EDC (included in the CarboxyLink Immobilization Kit), carboxyl-containing molecules become permanently attached to the support by stable amide bonds. Once a molecule is coupled, the prepared affinity column can be used multiple times in typical protein affinity purification procedures. CarboxyLink Coupling Resins can also be used to immobilize other kinds of molecules using alternative amine-reactive crosslinking chemistries.
Caution: For Research Use Only. Not for use in diagnostic procedures. Store at 4˚C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty