Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ Immobilized p-Aminophenyl Phosphoryl Choline Agarose, Thermo Scientific

Supplier: Thermo Fisher Scientific
Pierce
20307 20307
PI20307EA 609.23 USD InStock
CAPI20307 PI20307
Pierce™ Immobilized p-Aminophenyl Phosphoryl Choline Agarose, Thermo Scientific
Protein Purification Protein Purification Reagents
Thermo Scientific Pierce p-Amino Phosphoryl Choline Agarose enables purification of C-reactive protein (CRP) from plasma, ascites and other biological fluids.

  • Ligand: p-aminophenyl phosphoryl choline
  • Support: crosslinked 6% beaded agarose
  • Binding capacity: greater than 3 mg of human C-reactive protein per milliliter of settled resin
  • Format: 50% resin slurry in 0.02% sodium azide
  • Binding condition: Tris or borate-buffered saline (pH 8 to 8.5) with 1 to 2 mM calcium chloride
  • Elution condition: Tris or borate-buffered saline (pH 8 to 8.5) with 2 mM EDTA
  • Reusable: resin can be regenerated and reused several times

Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at 4˚C.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty