Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ SDS-Out™ Sodium Dodecyl Sulfate Precipitation Reagent, Thermo Scientific

Supplier: Thermo Scientific
SDS-Out™
20308 20308
PI20308EA 356.68 USD InStock
CAPI20308 PI20308
Pierce™ SDS-Out™ Sodium Dodecyl Sulfate Precipitation Reagent, Thermo Scientific
Cleaning Agents Decontaminants
SDS-Out™ SDS Precipitation Kit provides convenient removal of SDS from small-volume (0.1 to 10 mL) protein samples.

  • Convenient removal of SDS from microliter-volume samples
  • Used for samples containing 0.1 to 1% SDS
  • Minimal dilution of small-volume samples
  • Protein recovery is 90 to 100%

It has been tested with BSA, cytochrome c, soybean trypsin inhibitor, ovalbumin, ribonuclease A, myoglobin, and human IgG.

Delivery information: The kit includes SDS Precipitation reagent (10 mL), spin cup columns (12 columns) and collection tubes (12 tubes).

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty