Would you like to log in again?
Pierce™ Protein A Agarose, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
20334 20356 20333 20334 20333 20356
PI20334EA 1858.71 USD InStock
PI20334 PI20356 PI20333 CAPI20333 CAPI20334 CAPI20356
Pierce™ Protein A Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Protein A Agarose is a standard-capacity Protein A beaded agarose resin for use in a variety of antibody affinity purification methods.
  • Immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein A is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non-specific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilized Protein A
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty