Would you like to log in again?
Pierce™ Recombinant Protein A Plus Agarose, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
20365 20366 20365 20366
PI20365EA 500.68 USD InStock
PI20365 PI20366 CAPI20365 CAPI20366
Pierce™ Recombinant Protein A Plus Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ Recombinant Protein A agarose is a beaded agarose resin to which a genetically engineered form of Protein A has been immobilized for use in IgG purification applications where the native ligand has undesirable effects.
  • Immobilized Protein A is ideal for polyclonal IgG purification from human and rabbit
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Standard capacity beads with a normal load of immobilized Protein A
Although some non-essential regions have been removed, recombinant is similar to native Protein A in overall size and IgG-binding domains. The protein has four high affinity binding sites capable of specifically binding to the Fc region of immunoglobulin molecules from several species.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty