Would you like to log in again?
Pierce™ AminoLink™ Affinity Coupling Resin, Thermo Scientific
Supplier: Thermo Fisher Scientific
AminoLink™
20382 20381 20382 20381
PI20382EA 731.45 USD InStock
CAPI20381 CAPI20382 PI20382 PI20381
Pierce™ AminoLink™ Affinity Coupling Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups to enable covalent immobilization of antibodies and other proteins through primary amines.
  • Ideal for antibodies and other proteins – immobilize molecules via primary amines (-NH2)
  • Efficient (>85%) coupling over a wide range of pH (4 to 10) and buffer conditions (PBS or other non-amine buffer, with or without organic solvent)
  • Coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand
  • Better than immobilization to CNBr-activated agarose as the bond is more stable and uncharged, resulting in less non specific binding in affinity purification procedures
  • Prepared affinity resin is adaptable to column and batch affinity techniques and the resin is reusable for typical applications based on protein binding interactions
The AminoLink Resin immobilization reaction involves spontaneous formation of Schiff base bonds between aldehydes and amines and their subsequent stabilization by incubation with a mild reductant.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty