Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ Protein G Agarose, Thermo Scientific

Supplier: Thermo Fisher Scientific
Pierce
22851 20399 22852 20397 22851 20397 22852 20398 20398 20399
PI22852EA 1581.78 USD InStock
PI22852 PI20398 PI20397 PI22851 PI20399 CAPI22851 CAPI20397 CAPI20398 CAPI20399 CAPI22852
Pierce™ Protein G Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce Protein G and Protein G Plus Agarose is a premium-quality affinity resin for antibody purification.

  • Ideal for polyclonal IgG purification from mouse, human, cow, goat, and sheep serum, including human IgG3 and mouse IgG1 isotypes
  • Agarose resin support is crosslinked 6% beaded agarose (CL-6B)
  • Protein G made by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilized Protein G

Pierce Protein G Agarose consists of recombinant Protein G that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose (CL-6B). This particular variety of the affinity resin provides the most versatile combination of chromatographic features for high yield and high purity purification of whole IgG from mammalian serum samples. The agarose beads have physical and chemical properties suitable for many affinity purification systems.

Delivery information: Product is shipped at ambient temperature.

Caution: For Research Use Only. Not for use in diagnostic procedures. Store product at 4 °C.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty