Would you like to log in again?
Pierce™ Protein A/G and A/G Plus Agarose, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
20423 20422 20421 20423 20422 20421
PI20423EA 594.22 USD InStock
CAPI20423 PI20423 PI20422 PI20421 CAPI20421 CAPI20422
Pierce™ Protein A/G and A/G Plus Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ Protein A/G agarose and A/G Plus Agarose is a affinity resin for antibody purification.
  • Immobilized Protein A/G is ideal for polyclonal IgG purification from nearly any mammalian species
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein A/G is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilized Protein A/G
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty