Would you like to log in again?
Pierce™ Protein L Agarose, Thermo Scientific
Supplier: Thermo Fisher Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Pierce
20512 20510 20510 20512
PI20512EA 1637.45 USD InStock
CAPI20512 PI20512 PI20510 CAPI20510
Pierce™ Protein L Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce Protein L Agarose is a premium-quality affinity resin for antibody purification.
  • Immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein L is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilized Protein L
Pierce Protein L Agarose consists of recombinant Protein L that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose (CL-6B)
Pierce Protein L Agarose is a versatile affinity resin for purification of mammalian IgG isotypes that contain specific kappa light chains. Protein L is especially suited for isolating mouse IgG VkI and human IgG VkI, VkIII and VkIV monoclonal antibodies.
This particular variety of the resin provides the most versatile combination of chromatographic features for high yield and high purity purification of target immunoglobulins. The agarose beads have physical and chemical properties suitable for many affinity purification systems.
Delivery information: Product is shipped at ambient temperature.
Caution: For Research Use Only. Not for use in diagnostic procedures. Store product at 4-80˚C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty